Learn More
HYDRANAL™ - Methanol dry, Reagent for volumetric one-component KF titration (working medium), Honeywell Fluka™
Brand: Honeywell Fluka 34741-2.5L
Code : NEW
Additional Details : CAS Number : 67-56-1 Weight : 1.97500kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Specifications
HYDRANAL™-Methanol dry | |
9°C (48.2°F) | |
67-56-1 |
0.79g/mL | |
2.5L | |
UN number : 1230 |
Safety and Handling
- HYDRANAL-Methanol dry
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.