missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Formamide dry, Solvent for KF titration, Honeywell Fluka™

Product Code. 15654360 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15654360 6 x 1 L Pack of 6
15644360 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15654360 Supplier Honeywell-Hydranal Supplier No. 347246X1L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Solvent for Karl Fischer titration

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CH3NO
Molecular Weight (g/mol) 45.04
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
Synonym Formamide, Amide C1, Formic amide
IUPAC Name formamide
SMILES NC=O

Specifications

Melting Point 2°C to 3°C (literature)
Boiling Point 210°C
Density 1.134 g/mL at 25 °C(lit.)
CAS 68007-08-9, 75-12-7
Quantity 6 x 1 L
CAS Max % 100.0000%
Flash Point 175°C
Linear Formula HCONH2
Packaging Glass Bottle
Beilstein 505995
Grade Karl Fischer
Chemical Name or Material HYDRANAL™ - Formamide dry, Solvent for KF titration
Show More Show Less
compliance-icons
Product Identifier
  • HYDRANAL-Formamide dry
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: formamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.