missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - CRM Water Standard 10.0, Certified reference material for Karl Fischer titration (water content 10 mg/g = 1.0%), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 x 8 mL
Unit Size:
80mL
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™ - CRM Water Standard 10.0 |
| Name Note | Certified reference material for Karl Fischer titration (water content 10 mg/g = 1.0%) |
| Boiling Point | 109°C |
| CAS Min % | ≥20.0000% |
| CAS Max % | <30.0000%, <70.0000% |
| CAS | 71-36-3, 108-32-7 |
| Density | 1.000 g/cm3 |
| Quantity | 10 x 8 mL |
| Flash Point | 32°C |
| Assay Percent Notes | effective water volumetric |
| Show More |
Safety and Handling
Product Identifier
- HYDRANAL CRM Water Standard 10.0
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : propylene carbonate,o-xylene, butan-1-ol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction