missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat Oil, Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka™

Product Code. 15674620 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL
6 x 500 mL
Unit Size:
100mL
500mL
Pack of 6
This item is not returnable. View return policy

Product Code. 15674620

Brand: Honeywell-Hydranal 348686X100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anolyte for coulometric Karl Fischer titration in oils (chloroform-methanol-xylene-based), preferred for cells with diaphragm

Advantages of Hydranal Coulometric Reagents

  • Easy to use
  • High accuracy for trace amounts of water
  • Stable conditions of the titration vessel
  • Broad product range
  • Long shelf life (up to five years)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - Coulomat Oil
Name Note Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm
CAS Min % ≥20.0000%, ≥1.0000%, ≥20.0000%
CAS Max % <30.0000%, <5.0000%, <5.0000%, <20.0000%
CAS 67-66-3, 288-32-4, 95-47-6, 7446-09-5
Density 1.042 g/cm3
Quantity 6 x 100 mL
Flash Point 8°C
Packaging Glass Bottle
UN Number UN1992
Synonym Methyl alcohol
Grade Karl Fischer
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Coulomat Oil
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanol, o-xylene,trichloromethane; chloroform, imidazole,1H-Imidazole monohydriodide, sulphur di
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.