Learn More
HYDRANAL™ - Coulomat AK, Reagent for coulometric KF titration in ketones (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka
Brand: Honeywell Fluka 34820-500ML
160.14 GBP valid until 2024-12-31
Use promo code "21961" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.60000kg Transport : UN number : 1992 Chem class : 3 Pack group : III
Specifications
1.200 g/cm3 | |
500 mL | |
Reagent for coulometric KF titration in ketones (anolyte solution), preferred for cells with diaphragm | |
≥1.0000% | |
60°C | |
UN1992 |
32°C | |
HYDRANAL™ - Coulomat AK | |
109-86-4,67-66-3,75-89-8,101023-55-6,68007-08-9,7446-09-5 | |
<5.0000% | |
Glass Bottle | |
Karl Fischer |
Safety and Handling
- HYDRANAL
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H351-Suspected of causing cancer.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 2-methoxyethanol; ethylene glycol monomethyl ether, trichloromethane; chloroform, 2,2,2-Trifluoroeth