missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Coulomat AG, Reagent for coulometric KF titration (anolyte solution), for cells with and without diaphragm, Honeywell Fluka™

Product Code. 15634500 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
6 x 500 mL
6 x 1 L
Unit Size:
1 L
250mL
500mL
Pack of 6
This item is not returnable. View return policy

Product Code. 15634500

Brand: Honeywell Fluka 348366X1L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material HYDRANAL™-Coulomat AG
CAS 7446-09-5, 68007-08-9, 111-42-2, 67-56-1, 288-32-4
Quantity 6 x 1 L
Flash Point 14°C (57.2°F)
UN Number UN1230
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • hydranal coulomat AG
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H311-Toxic in contact with skin.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : methanol, 2,2'-iminodiethanol; diethanolamine, imidazole, sulphur dioxide, 1H-imidazole monohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.