missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 2, Honeywell Fluka™

Product Code. 16676163 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
6 x 1 L
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16676163 6 x 1 L Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16676163 Supplier Honeywell Fluka Supplier No. 348066X1LSC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HYDRANAL™ - Composite 2, Titrating agt for vlmtrc 1-comp Karl Fischer titration (methnl free)

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - Composite 2
CAS Max % <90%
Color Brown
Density 1.110 g/cm3 (20°C)
pH 3 to 4
CAS 7446-09-5
Quantity 6 x 1 L
Flash Point 103°C
Synonym DE Solvent; 2-(2-Ethoxyethoxy)ethanol; CARBITOL; Diethylene glycol ethyl ether; Ethyldiglycol
Grade Volumetric One-Component KF Titration
compliance-icons
Product Identifier
  • HYDRANAL NEXTGEN Composite 2 SC
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-(2-Ethoxyethoxy)ethanol, sulphur dioxide, Imidazole, iodine, 2-methylimidazole, 1H-Imidazole monoh
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.