missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Composite 1, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™

Brand:  Honeywell Fluka 34827-6X1L

Code : NEW

Additional Details : CAS Number : 111-90-0 Weight : 6.66000kg


 View more versions of this product

Product Code. 15644490

  • £609.91 / Pack of 6
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

90°C (194°F)
6 x 1 L
HYDRANAL™-Composite 1
1.11g/mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL-Composite 1

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H351-Suspected of causing cancer.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

Supplemental information
  • MIXTURE LIST-Contains : 2-(2-Ethoxyethoxy)ethanol, imidazole, 2-Methylimidazole, sulphur dioxide, iodine, 1H-Imidazole monoh

SDS Documents

Documents

Certificates
Special Offers

Special Offers