missing translation for 'onlineSavingsMsg'
Learn More

Hydralazine Lactosone Ring-opened Adduct, TRC

Product Code. 30402144
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402144 25 mg 25mg
30394638 50 mg 50mg
30400087 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402144 Supplier Toronto Research Chemicals Supplier No. TRCH67515025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydralazine Lactosone Ring-opened Adduct
Analyte or Component Names Hydralazine Lactosone Ring-opened Adduct
Purity Grade Notes HPLC
Shipping Condition Room Temperature
Molecular Formula C20H26N4O10
InChI Formula InChI=1S/C20H26N4O10/c25-6-10(27)17(34-20-16(32)13(29)12(28)11(7-26)33-20)14(30)15(31)19-23-22-18-9-4-2-1-3-8(9)5-21-24(18)19/h1-5,10-17,20,25-32H,6-7H2/t10?,11-,12+,13+,14-,15-,16-,17-,20+/m1/s1
Solution Type Neat
SMILES OC[C@H]1O[C@@H](O[C@H](C(CO)O)[C@H](O)[C@@H](O)C2=NN=C3N2N=CC4=C3C=CC=C4)[C@H](O)[C@@H](O)[C@H]1O
Industry Type Pharmaceutical
Molecular Weight (g/mol) 482.44
Quantity 25 mg
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.