missing translation for 'onlineSavingsMsg'
Learn More

Hydralazine Hydrochloride, TRC

Product Code. 30138493
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138493 1 g 1g
30132801 50 mg 50mg
18052720 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138493 Supplier Toronto Research Chemicals Supplier No. TRCH7165311G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304-20-1
Molecular Formula C8 H8 N4 . Cl H
Molecular Weight (g/mol) 196.64
Synonym Hydralazine hydrochloride, 1-Hydrazinophthalazine hydrochloride, 1(2H)-Phthalazinone, hydrazone, monohydrochloride, Phthalazine, 1-hydrazino-, monohydrochloride, (Phthalazin-1-yl)hydrazine hydrochloride, 1-Hydrazinophthalazine hydrochloride, 1-Hydrazinophthalazine monohydrochloride, Apresoline hydrochloride, Apulon, Hydralazine chloride, Hydralazine hydrochloride, Hydralazine monohydrochloride, Lopres
IUPAC Name phthalazin-1-ylhydrazine;hydrochloride

Specifications

Quantity 1 g
InChI Formula InChI=1S/C8H8N4.ClH/c9-11-8-7-4-2-1-3-6(7)5-10-12-8;/h1-5H,9H2,(H,11,12);1H
SMILES Cl.NNc1nncc2ccccc12
Formula Weight 196.0516
Percent Purity >95%
Chemical Name or Material Hydralazine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.