missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ composite 5, One component volumetric reagents, J.T. Baker™

Product Code. 10377541 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10377541 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10377541 Supplier JT Baker Supplier No. 8890.2500
  • ONLINE EXCLUSIVE PRICE
    £726.33
    £431.89 / 2.5 L
    Save £294.44
    40% Off
 Restricted Item
Estimated Shipment: 24-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Pyridine free one-component reagent for volumetric Karl Fischer titration

  • Volumetric reagents for water determination by Karl Fischer titration
  • No matter what your requirements are for volumetric systems, HYDRA-POINT™ Karl Fischer reagents make water determination easy and trouble-free
  • They are pyridine-free and specially formulated for existing application procedures to give you consistent, reliable results for all types of samples
  • Formulated to reach each endpoint quickly, accurately and with reproducibility
  • Accuracy- reagents have titres that are more stable with less drift, helping you save time and reduce reagent usage
  • To be used with HYDRA-POINT™ Methanol dry product no. 8898

Specifications

Quantity 2.5 L
UN Number 2922
Concentration or Composition (by Analyte or Components) 5 mg/mL H2O
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.