missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRA-POINT™ Buffer Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Description
- For Karl Fischer titration
Specifications
Specifications
| Chemical Name or Material | HYDRA-POINT™ Buffer Solution |
| Quantity | 1 L |
| Flash Point | 11.00°C |
| Packaging | Narrow Mouth Amber Glass Bottle |
| Grade | BAKER ANALYZED™ Reagent |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H361-Suspected of damaging fertility or the unborn child.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction