missing translation for 'onlineSavingsMsg'
Learn More

Hydantoin-5-acetic acid, 98%

Product Code. 11489253
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489253 25 g 25g
11479253 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489253 Supplier Thermo Scientific Alfa Aesar Supplier No. A11731.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 5427-26-9 | C5H6N2O4 | 158.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5427-26-9
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.11
MDL Number MFCD00005267
InChI Key DQQLZADYSWBCOX-UHFFFAOYNA-N
Synonym hydantoin-5-acetic acid, 5-hydantoinacetic acid, 2,5-dioxoimidazolidin-4-yl acetic acid, 5-hydantoin acetic acid, 2-2,5-dioxoimidazolidin-4-yl acetic acid, 4-imidazolidineacetic acid, 2,5-dioxo, 2,5-dioxo-1,3-diazolidin-4-ylacetic acid, 2-2,5-dioxo-1,3-diazolidin-4-yl acetic acid, 5-carboxymethylhydantoin, acmc-20akr6
PubChem CID 95492
IUPAC Name 2-(2,5-dioxoimidazolidin-4-yl)acetic acid
SMILES OC(=O)CC1NC(=O)NC1=O

Specifications

Melting Point ∼217°C (decomposition)
Quantity 25 g
Beilstein 83035
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material Hydantoin-5-acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.