Learn More
Ansell™ Hycron™ 27-805 Series Blue Nitrile Heavy Weight Gloves
Product Code. p-8005279 Shop All Ansell Products
Click to view available options
Size:
9
10
11
Unit Size:
12 pairs
Product Code. 12630802
Brand: Ansell™ 27805110
Only null left
Add to basket
Add to basket
Three times more durable than heavy-duty leather. Unique Ansell nitrile formulation offers superior resistance to sharp and abrasive materials.
  • Nitrile-dipped coating is stronger and more flexible than PVC.
  • Provides excellent barrier to grease and oil
  • Does not degrade like leather or cotton.
  • Made without silicone.
  • Guaranteed clean for metals and motor manufacture.
  • Cut and sewn construction
  • Cotton jersey liner
  • Fully-coated finishing
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Certifications/Compliance Standards CATEGORY II, EN 388 : 4221
Cuff Material Nitrile
Coating Material Nitrile
Color Green
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Heavyweight Gloves
Size 11
Glove Cuff Style Long Safety
Liner Material Cotton Jersey
Lining Lined
Palm Type Fully coated-palm
Quantity 144 Pairs
Resistant To Sharp and abrasive materials
Series 27-805