missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 27-600 Nitrile Coated Heavy Duty Work Gloves - Safety Cuff

Product Code. 12610802 Shop All Ansell Products
Change view
Click to view available options
Size:
8
9
10
11
Unit Size:
12 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12610802 9 12 pairs
12600802 8 12 pairs
12680792 10 12 pairs
12690792 11 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12610802 Supplier Ansell Supplier No. 27607090
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Gloves feature a unique coating formulation that provides increased resistance against sharp and abrasive environments, in addition to oil and grease.

  • Designed to be extra tough for use in environments that involve rough, abrasive materials
  • Constructed of cotton and jersey fabric for a high level of user comfort
  • Unique nitrile coating provides barrier protection against oil and grease
  • Safety cuff increases protection to the wrist
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Cut and Sewn
Coating Material Nitrile
Color Blue, Natural
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 9
EN388 Standard 4121B
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, and Metal Fabrication
Glove Cuff Style Safety
Series ActivArmr™ 27-607
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.