missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Water for Injection (WFI) Quality Water

Product Code. 10317002
Change view
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
200 L
Unit Size:
10 L
1000mL
20 L
200 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10317002 1 L 1000mL
10464292 500 mL 500mL
11531861 10 L 10 L
11541861 20 L 20 L
12329882 200 L 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10317002 Supplier Cytiva Supplier No. SH30221.10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Supports cell culture growth and biomanufacturing process.

  • Format: Liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym dihydrogen oxide, dihydrogen monoxide
SMILES O

Specifications

IUPAC Name water
Molecular Weight (g/mol) 18.02
Quantity 1 L
Chemical Name or Material Water
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.