missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Adult Bovine Serum, New Zealand Origin

Product Code. 16665765
Change view
Click to view available options
Quantity:
500 mL
1000 mL
3000 mL
Processing:
Irradiated
Non-Irradiated
Unit Size:
1 L
3 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing Unit Size
16665765 3000 mL Non-Irradiated 3 L
16635765 1000 mL Irradiated 1 L
16655765 3000 mL Irradiated 3 L
16625765 500 mL Non-Irradiated 500mL
16645765 1000 mL Non-Irradiated 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16665765 Supplier Cytiva Supplier No. SH30528.05NI

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA), average age at time of collection is 6 to 8 years

TRUSTED_SUSTAINABILITY

Specifications

Age 6 to 8 years
Country of Origin New Zealand
Product Type Adult Bovine Serum
Quantity 3000 mL
Sterilization Method Triple-filtered
Processing Non-Irradiated
Product Line HyClone
Test Methods Tested According to European Pharmacopoeia Protocols
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.