missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Adult Bovine Serum, New Zealand Origin

Product Code. 16635765
Change view
Click to view available options
Quantity:
500 mL
1000 mL
3000 mL
Processing:
Irradiated
Non-Irradiated
Unit Size:
1 L
3 L
500mL
Product Code. Quantity Processing unitSize
16635765 1000 mL Irradiated 1 L
16655765 3000 mL Irradiated 3 L
16625765 500 mL Non-Irradiated 500mL
16645765 1000 mL Non-Irradiated 1 L
16665765 3000 mL Non-Irradiated 3 L
5 options
This item is not returnable. View return policy

Product Code. 16635765

Brand: Cytiva SH30528.03

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA), average age at time of collection is 6 to 8 years

TRUSTED_SUSTAINABILITY

Specifications

Age 6 to 8 years
Country of Origin New Zealand
Product Type Adult Bovine Serum
Quantity 1000 mL
Sterilization Method Triple-filtered
Processing Irradiated
Product Line HyClone
Test Methods Tested According to European Pharmacopoeia Protocols
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.