missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Tetracycline Screened Characterized Fetal Bovine Serum, US Origin
Shop All Cytiva Products
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
Tetracycline Screened FBS is designed for researchers using tetracycline-regulated gene expression systems in cultured cells and is suitable for expression studies in Tet-on/Tet-off systems.
Specifications
Specifications
| Country of Origin | United States |
| Form | Liquid |
| Specialty Type | Tetracycline screened |
| Quantity | 500 mL |
| Processing | None |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Heat Inactivated | No |
| Hemoglobin Concentration | ≤ 10 mg/dL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction