missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ SFM4MegaVir Powdered Media

Product Code. 10123442
Click to view available options
Quantity:
5 L
Unit Size:
5 L
This item is not returnable. View return policy

Product Code. 10123442

Brand: Cytiva SH30587.01

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Increase process yields in the manufacture of viral vaccines

HyClone SFM4MegaVir is a regulatory-friendly and animal-derived component-free (ADCF) cell culture medium. The medium was designed to increase process yields in the manufacture of viral vaccines in a variety of anchorage dependent cell lines including Vero, MDCK, COS-7, and MDBK cells.

SFM4MegaVir has been successfully tested in a variety of culture systems for cell growth and virus production including T-flasks and suspension microcarrier cultures.

SFM4MegaVir is available in liquid and powder formats in user-friendly packaging.

  • ADCF cell culture medium
  • Designed for high viral vaccine production
  • Suitable for direct adaptation
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Sterility Non-sterile
With Additives Phenol Red
Without Additives No Glutamine, No Sodium Bicarbonate
Quantity 5 L
Content And Storage 2 to 8°C, away from light and moisture
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.