missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ SFM4CHO™ Media
Description
- Formulated using proprietary lipid complexing process for enhanced stability and growth promotion of various CHO cell lines
Specifications
Specifications
| Form | Liquid |
| Product Type | SFM4CHO Liquid Media |
| Sterilization Method | 0.2 μm Sterile-filtered |
| With Additives | Sodium Bicarbonate, Poloxamer 188 |
| Without Additives | No Glutamine, No Phenol Red, No HEPES |
| Quantity | 1000 mL |
| Certifications/Compliance | cGMP |
| Content And Storage | 2° to 8°C, away from light |
| Description | SFM4CHO media |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?