missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Research Grade Fetal Bovine Serum, South American Origin

Product Code. 12330273
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12330273 100 mL 100mL
12336913 50 mL 50mL
16635805 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12330273 Supplier Cytiva Supplier No. SV30160.02HI
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Strict manufacturing and quality control procedures ensure quality is suitability for research applications

  • Filtered through three sequential 100 nm pore-size rated filters
  • Full traceability compliant with current Good Manufacturing Practices (cGMP)
  • Extensive quality, performance, and analytical testing
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
  • Aseptically packaged in a Class 100 cleanroom
  • Pooled after filtration and before final packaging
  • Each lot of serum is tested to ensure quality and sterility
  • Batch testing available on request
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Complies with EU Regulations (Directive 92/118/EEC)
Country of Origin South America
Product Type Fetal Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Grade Research Grade
Processing Heat Inactivated
Volume (Metric) 100 mL
For Use With (Application) Research
Heat Inactivated Yes
Product Line HyClone
Shipping Condition Frozen
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.