missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Porcine Serum, New Zealand Origin

Product Code. 15154732
Change view
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15154732 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15154732 Supplier Cytiva Supplier No. SH30908.04HI

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enhances Cell Culture Growth

TRUSTED_SUSTAINABILITY

Specifications

Country of Origin New Zealand
Product Type Porcine Serum
Quantity 1000 mL
Sterilization Method Sequential Triple-filtered, 200 nm
Processing Irradiated, Heat Inactivated
Endotoxin Concentration ≤ 100 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤ 50 mg/dL
Product Line HyClone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.