missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Calf Serum (U.S.)
Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.
Brand: Cytiva SH30118.03IR25-40
Additional Details : Weight : 0.50000kg
Description
- Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old
Specifications
Newborn Calf Serum (U.S.) | |
500 mL | |
Irradiated, 25 to 40 doses | |
No |