missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Newborn Calf Serum (U.S.)
Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.
Brand: Cytiva SH30118.04
Code : 85
Additional Details : Weight : 0.01000kg
Description
- Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old
Specifications
Albumin, Iron, Lactate Dehydrogenase, Osmolality, Chloride, pH, Gamma Globulin, Glucose, IgG, Sodium, TIBC, Alkaline Phosphatase, Total Bilirubin, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus | |
1000 mL | |
Sequential Triple-filtered, 100 nm |
Newborn Calf Serum (U.S.) | |
Sterile | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |