missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Bovine Serum, New Zealand Origin

Product Code. 16675775
Change view
Click to view available options
Processing:
Heat Inactivated
Non-Irradiated
None
Unit Size:
1 L
Product Code. Processing unitSize
16675775 Heat Inactivated 1 L
16685775 Non-Irradiated 1 L
16665775 None 1 L
3 options
This item is not returnable. View return policy

Product Code. 16675775

Brand: Cytiva SH30560.02HI

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)

  • Carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases
TRUSTED_SUSTAINABILITY

Specifications

Age <10 days
Country of Origin New Zealand
Product Type Newborn Calf Bovine Serum
Quantity 1000 mL
Sterilization Method Sequential Triple-filtered, 100 nm
Processing Heat Inactivated
Endotoxin Concentration ≤ 25 EU/mL
Hemoglobin Concentration ≤ 25 mg/dL
Product Line HyClone
Test Methods European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.