missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Newborn Bovine Calf Serum

Product Code. 17692075
Change view
Click to view available options
Quantity:
25 to 40 Doses
30 to 45 Doses
Unit Size:
1 L
Product Code. Quantity unitSize
17692075 30 to 45 Doses 1 L
17682075 25 to 40 Doses 1 L
2 options
This item is not returnable. View return policy

Product Code. 17692075

Brand: Cytiva SH30401.02IH3045

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).

  • Supplement cell culture growth with New Zealand sourced Cytiva HyClone Newborn Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
    • Sourced from animals that are typically less than 10 days old
    • Traceable
    • Conforms to USDA regulations
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -10°C or Lower, Once Thawed Store at 2°C to 8°C
Country of Origin New Zealand
Product Type Newborn Calf Bovine Serum
Quantity 30 to 45 Doses
Volume (Metric) 1000 mL
For Use With (Application) Enhance Growth of Cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.