missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Minimal Essential Medium Variations: Powder

Product Code. 11501791
Click to view available options
Quantity:
2 x 5 L
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 11501791

Brand: Cytiva SH30008.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Supports cell growth in research and biomanufacturing cell culture processes.

  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage 2° to 8°C, away from light and moisture
    Form Powder
    Sterility Non-sterile
    With Additives Glutamine, None
    Without Additives No Sodium Bicarbonate
    Quantity 2 x 5 L
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.