missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Iron-Supplemented Newborn Calf Serum, New Zealand Origin

Product Code. 16655785
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Irradiated
Irradiated (30 to 40 kGy)
None
Unit Size:
1 L
100mL
500mL
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing unitSize
16655785 100 mL Irradiated 100mL
16665785 500 mL Irradiated (30 to 40 kGy) 500mL
16675785 1000 mL Irradiated (30 to 40 kGy) 1 L
11572830 - None Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16655785 Supplier Cytiva Supplier No. SH30626.01IR

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Contains three to four times as much available iron and transferrin as FBS or equine serum

  • Carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases
  • Contains three to four times as much available iron and transferrin as FBS or equine serums
  • Collected from cattle that are typically less than 10 days old
TRUSTED_SUSTAINABILITY

Specifications

Age <10 days
Country of Origin New Zealand
Product Type Newborn Calf Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Processing Irradiated
Iron Supplemented Yes
Product Line HyClone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.