missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum, EMEA Tested
Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)
£133.00 - £139.74
Specifications
Age | 16 to 22 weeks |
---|---|
Product Type | Calf Bovine Serum |
Endotoxin Concentration | ≤ 10 EU/mL |
Hemoglobin Concentration | ≤ 10 mg/dL |
Iron Supplemented | Yes |
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
11987328
|
Cytiva
SH30534.04 |
Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16615775
|
Cytiva
SH30534.04NI |
Non-Irradiated |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Contains three to four times as much available iron and transferrin as FBS or equine serum
Specifications
16 to 22 weeks | |
≤ 10 EU/mL | |
Yes | |
European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests |
Calf Bovine Serum | |
≤ 10 mg/dL | |
HyClone™ |