missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Hank's 1X Balanced Salt Solutions

Product Code. 10452372
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1000 mL
With Additives:
Calcium, Magnesium
Calcium, Magnesium, Phenol Red
Phenol Red
Phenol Red, Calcium, Magnesium
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
No Phenol Red
Unit Size:
1000mL
100mL
500mL
Pack of 6
This item is not returnable. View return policy

Product Code. 10452372

Brand: Cytiva SH30268.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Buffer cell culture growth with 1X balanced salt solutions

  • Used for washing and treating tissues and cells
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Content And Storage 15° to 30°C
Concentration 1 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
Sterilization Method 0.1 μm Sterile-filtered
With Additives Calcium, Magnesium
Without Additives No Phenol Red
Endotoxin Level =1.0EU/mL
pH 7.0 to 7.4
Quantity 1000 mL
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.