missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Hank's 1X Balanced Salt Solutions

Product Code. 10227993
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
6 x 500 mL
6 x 1000 mL
With Additives:
Calcium, Magnesium
Calcium, Magnesium, Phenol Red
Phenol Red
Phenol Red, Calcium, Magnesium
Without Additives:
No Calcium, No Magnesium
No Calcium, No Magnesium, No Phenol Red
No Phenol Red
Unit Size:
1000mL
100mL
500mL
Pack of 6
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity With Additives Without Additives unitSize
10227993 6 x 1000 mL Calcium, Magnesium No Phenol Red Pack of 6
11546291 1000 mL - No Calcium, No Magnesium, No Phenol Red 1000mL
10553764 6 x 500 mL Phenol Red No Calcium, No Magnesium Pack of 6
10739674 500 mL Calcium, Magnesium No Phenol Red 500mL
10041432 1000 mL Phenol Red No Calcium, No Magnesium 1000mL
10532003 500 mL Phenol Red No Calcium, No Magnesium 500mL
10601993 1000 mL Phenol Red, Calcium, Magnesium - 1000mL
10617394 500 mL - No Calcium, No Magnesium, No Phenol Red 500mL
10309473 100 mL Phenol Red No Calcium, No Magnesium 100mL
10225362 500 mL Calcium, Magnesium, Phenol Red - 500mL
10452372 1000 mL Calcium, Magnesium No Phenol Red 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10227993 Supplier Cytiva Supplier No. SH30268.LS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Buffer cell culture growth with 1X balanced salt solutions

  • Used for washing and treating tissues and cells
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Content And Storage 15° to 30°C
Concentration 1 X
Form Liquid
Product Type Hanks' Balanced Salt Solution
Sterilization Method 0.1 μm Sterile-filtered
With Additives Calcium, Magnesium
Without Additives No Phenol Red
Endotoxin Level =1.0EU/mL
pH 7.0 to 7.4
Quantity 6 x 1000 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.