missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ G418 Sulfate

Product Code. 11596471
Click to view available options
Quantity:
20 mL (50 mg/mL)
5 g
Unit Size:
20mL
5g
This item is not returnable. View return policy

Product Code. 11596471

Brand: HyClone products Cytiva SV30069.01

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Control or destroy destructive microorganisms with HyClone™ G418 Sulfate.

  • Aminoglycoside is used as a selective agent of transfected mammalian, yeast, plant and bacteria cells
  • Related to Gentamycin
  • Interferes with the function of 80S ribosomes and protein synthesis in eukaryotic cells
  • Resistance is conferred by the bacterial gene for aminoglycoside-3'-phosphotransferase that can be expressed in eukaryotic cells
  • Common working concentration for G418 is 300 to 1000μg/mL
TRUSTED_SUSTAINABILITY

Specifications

Description G418 Sulfate Solution, 50mg/mL
Antibiotic Spectrum Selection agent
Concentration 50 mg/mL
Form Liquid
Purity or Quality Grade >9%
Quantity 20 mL (50 mg/mL)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.