missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ G418 Sulfate

Product Code. 11586471
Change view
Click to view available options
Quantity:
20 mL (50 mg/mL)
5 g
Unit Size:
20mL
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11586471 5 g 5g
11596471 20 mL (50 mg/mL) 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11586471 Supplier HyClone products Cytiva Supplier No. SV30068.02

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Control or destroy destructive microorganisms with HyClone™ G418 Sulfate.

  • Aminoglycoside is used as a selective agent of transfected mammalian, yeast, plant and bacteria cells
  • Related to Gentamycin
  • Interferes with the function of 80S ribosomes and protein synthesis in eukaryotic cells
  • Resistance is conferred by the bacterial gene for aminoglycoside-3'-phosphotransferase that can be expressed in eukaryotic cells
  • Common working concentration for G418 is 300 to 1000μg/mL
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 8°C
Description G418 Sulfate, >90% Purity
Antibiotic Spectrum Selection agent
Concentration 5 mg/mL
Form Powder
Purity or Quality Grade >90%
Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.