missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum, USDA Tested
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
None
Unit Size:
100mL
500mL
Description
Supplement cell culture growth with USDA-tested HyClone™ Fetal Bovine Serum (FBS), sourced from countries recognized as being free of BSE and FMD by the USDA.
Supplement cell culture growth with USDA-tested HyClone™ Fetal Bovine Serum (FBS), sourced from countries recognized as being free of BSE and FMD by the USDA.
Specifications
Specifications
| Additional Assays | Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus, Lactate Dehydrogenase, Osmolality, pH, Potassium, SGOT, SGPT, Sodium, Total Bilirubin, Total Protein |
| Certifications/Compliance | USDA Regulations |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
| Description | USDA Tested Fetal Bovine Serum, Central American Sourced |
| Country of Origin | United States |
| Product Type | Fetal Bovine Serum |
| Quantity | 100 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm |
| Grade | Characterized |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction