missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Fetal Bovine Serum (U.S.), Super Low IgG

Product Code. 10329953
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated, 25 to 40 doses
None
Unit Size:
100mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing unitSize
10329953 500 mL None 500mL
16221122 500 mL Irradiated, 25 to 40 doses 500mL
11547310 100 mL None 100mL
12360143 500 mL Heat Inactivated 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10329953 Supplier Cytiva Supplier No. SH30898.03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Processed using protein G chromatography to reduce the levels to those required for monoclonal antibody production

Fetal bovine serum (FBS) has natural levels of IgG that are too high for some cell culture applications. Lots can be prescreened to find the lowest naturally occuring levels, but as a biological product, FBS is frequently in short supply. Even lots with naturally low IgG levels may still contain too much for some applications. Examples of typical IgG levels in bovine serum products can be found in the table below.

  • Performance is equivalent to non-processed FBS
  • Suitable for applications where low levels of specific antibody are required (e.g. propagation of bovine viruses, monoclonal antibody production)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 8°C
Quantity 500 mL
Processing None
Heat Inactivated No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.