missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum (U.S.), Standard
Supplement cell culture growth with U.S. sourced quality cell culture-grade Cytiva HyClone™ Standard Fetal Bovine Serum (FBS)
Brand: Cytiva SH30088.03IR
Code : 85
Additional Details : Weight : 0.50000kg
Description
- Support cell culture growth
- Standard grade
- Aseptically packaged in Class 100 cleanroom
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
United States | |
500 mL | |
Sequential-filtered, 100 nm | |
Gamma Irradiated | |
For Information Only | |
For Information Only | |
USDA Inspected |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Standard Fetal Bovine Serum, US Origin | |
Fetal Bovine Serum | |
Sterile | |
Standard | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
No |