missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Fetal Bovine Serum (U.S.), Embryonic Stem (ES) Cell Screened

Product Code. 12339792
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Embryonic Stem Cell Screened
Heat Inactivated, ES Screened
Unit Size:
100mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Processing unitSize
12339792 500 mL Heat Inactivated, ES Screened 500mL
11501831 100 mL Embryonic Stem Cell Screened 100mL
10117272 500 mL Embryonic Stem Cell Screened 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12339792 Supplier Cytiva Supplier No. SH30070.03EH

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Augment growth of cells with U.S.-sourced HyClone™ Embryonic Stem (ES) Cell Screened Fetal Bovine Serum (FBS), filtered through serial 40nm pore-size rated filters and tested for endotoxin, hemoglobin, and extensive biochemical profile assays.

  • Filtered through sequential 40 nm (0.04μm) pore-size rated filters
  • Endotoxin: ≤ 10 EU/mL; Hemoglobin ≤ 10mg/dL
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays 1, 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1, B12, B2, B6, Biotin, Blood Urea Nitrogen, Calcium, Carotene, Chloride, Cholesterol, Copper, Creatinine, Folate, Gamma Globulin, Glucose, HDL, IgG, Inorganic Phosphorus, Insulin, Iron, Lactate Dehydrogenase, LDL, Magnesium, Niacin, Osmolality, Percent Saturation, pH, Phospholipids, Potassium, Progesterone, Retinol, Selenium, SGOT, SGPT, Sodium, TIBC, Tocopherol, Total Bilirubin, Total Protein, Triglycerides, Vitamin C
Certifications/Compliance EMEA Certified
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Country of Origin United States
Specialty Type ES cell screened
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 40 nm
Grade Defined, Specialty screened
Processing Heat Inactivated, ES Screened
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated Yes
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Yes
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.