missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum, Super Low IgG
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| Country of Origin | United States |
| Form | Serum |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Grade | Super Low IgG |
| Processing | Heat Inactivated, Irradiated (25 to 40 kGy) |
| Volume (Metric) | 500 mL |
| For Use With (Application) | Enhanceing Cell Culture Growth |
| Heat Inactivated | Yes |
| IgG levels | Super Low |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction