missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand)

Product Code. 12624247
Click to view available options
Quantity:
500 mL
1 L
Processing:
Heat Inactivated
Irradiated
Irradiated (25 to 35 doses)
Irradiated (30 to 40 doses)
None
missing translation for 'unitSize'
1 L
1000mL
500mL
This item is not returnable. View return policy

Product Code. 12624247

missing translation for 'mfr': Cytiva SH30406.03IR

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enhance growth of cells with New Zealand sourced HyClone™ Characterized Fetal Bovine Serum.

  • HyClone Characterized Fetal Bovine Serum (FBS), New Zealand Origin is carefully collected, processed, and filtered in our own New Zealand facility.
  • The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
  • One of the benefits of this isolation is that New Zealand has the fewest reported bovine diseases in the world, making Characterized FBS, New Zealand Origin an excellent source of bovine serum as it offers greater security.
  • Characterized FBS, New Zealand Origin is filtered through three sequential 100 nm (0.1 μm) pore size-rated filters.
  • Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles (true pool technology).
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Progesterone, Chloride, Insulin, Inorganic Phosphorus, PTH, Glucose, Cholesterol, Copper, HDL, Magnesium, LDL, Selenium, Triglycerides, IgG, Gamma Globulin, pH, Phospholipids, Osmolality, Alkaline Phosphatase, Iron, Lactate Dehydrogenase, TIBC, SGPT, Percent Saturation, SGOT, B1, B2, B6, Niacin, Biotin, Retinol, Carotene, Tocopherol, B12, Folate, Vitamin C, 1,25 Dihydroxy Vitamin D, Total Protein, Albumin, and Blood Urea Nitrogen
Content And Storage Store at or below -10°C
Country of Origin New Zealand
Form Solution
Product Type Fetal Bovine Serum
Quantity 1 L
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Grade Characterized
Processing Irradiated
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 20 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 25 mg/dL
Product Line HyClone
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.