missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Fetal Bovine Serum, Embryonic Stem (ES) Cell Screened
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- Serum suitability is critical for helping cells maintain their valuable undifferentiated state
- Screening includes plating efficiency, colony morphology, and toxicity tests
- Tested according to a protocol adapted from Gene Targeting, A.L. Joyner, 1995
Specifications
Specifications
| Country of Origin | United States |
| Quantity | 100 mL |
| Sterilization Method | Sequential-filtered, 40 nm |
| Volume (Metric) | 100 mL |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Heat Inactivated | Yes |
| Hemoglobin Concentration | ≤ 10 mg/dL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction