missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)
Click to view available options
Quantity:
500 mL
1 L
Processing:
HI, Irr - 25 to 40 doses
HI, Irr - 30 to 45 doses
Heat Inactivated
Irradiated (25 to 35 doses)
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
Unit Size:
1 L
500mL
Description
- Filtered through 3 sequential 0.1 μm pore size filters
Specifications
Specifications
| Additional Assays | Alkaline Phosphate, Lactate Dehydrogenase, SGPT, SGOT, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Glucose, Iron, TIBC, Percent Saturation, Gamma Globulin, IgG, pH, Osmolality |
| Certifications/Compliance | AQIS |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
| Country of Origin | Australia |
| Form | Liquid |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential Triple-filtered, 100 nm |
| Grade | Characterized |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction