missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)

Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.

Brand:  Cytiva SH30084.03IR25-40

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 16201102

  • £1160.00 / 500mL
Estimated Shipment: 12-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Filtered through 3 sequential 0.1 μm pore size filters
Specifications

Specifications

Alkaline Phosphate, Lactate Dehydrogenase, SGPT, SGOT, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Glucose, Iron, TIBC, Percent Saturation, Gamma Globulin, IgG, pH, Osmolality
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Fetal Bovine Serum
Sterile
Characterized
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
HyClone™
AQIS
Australia
500 mL
Sequential Triple-filtered, 100 nm
Irradiated (25 to 40 doses)
No
USDA Inspected
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers