missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ ES Screened Characterized Fetal Bovine Serum, US Origin
Click to view available options
Quantity:
100 mL
500 mL
Heat Inactivated:
No
Yes
Unit Size:
100mL
500mL
Description
ES Screened FBS supports the growth of undifferentiated murine embryonic stem cells. The screening includes plating efficiency, colony morphology, and toxicity tests.
Specifications
Specifications
| Country of Origin | United States |
| Form | Liquid |
| Specialty Type | ES cell screened |
| Quantity | 100 mL |
| Sterilization Method | 40 nm filtration |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Heat Inactivated | No |
| Hemoglobin Concentration | ≤ 10 mg/dL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction