missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Solution

Buffer cell culture with PBS in solution.

Brand:  Cytiva SH30028.04

Code : A0

Additional Details : CAS Number : 53123-88-9 Weight : 20.00000kg

 View more versions of this product

Product Code. 11586511

  • £331.00 / 20 L
Estimated Shipment: 23-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Format: Liquid
Specifications

Specifications

Manufactured under cGMP and ISO 9001 certified facilities
HyClone Dulbecco's Phosphate Buffered Salines (DPBS) Without Calcium, Magnesium, Phenol Red
Liquid
Sterile
None
≤0.025EU/mL
276±10mOsm/kg H2O
20 L
15° to 30°C
1 X
Dulbecco's Phosphate Buffered Saline
0.1 μm Sterile-filtered
No Calcium, No Magnesium
Serves as an irrigating, transporting or diluting fluid while maintaining cell tonicity and viability for a limited period of time
7 to 7.6
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers