missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Dulbecco's Phosphate Buffered Saline, Powder

Brand:  Cytiva SH30013.04

51.20 GBP valid until 2024-12-31
Use promo code "23281" to get your promotional price.



Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10708334

  • £58.80 / 50 L
Estimated Shipment: 21-11-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Buffer cell culture with HyClone™ Dulbecco's Phosphate Buffered Saline (PBS), Powder.

Specifications

Specifications

Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Without Calcium, Magnesium (Powder), 50L
Non-sterile
50 L
2° to 8° C
Powder
No Calcium, No Magnesium
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers