missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Donor Adult Bovine Serum (U.S.)

Product Code. 16635745
Click to view available options
Quantity:
1 L
Processing:
Irradiated
Irradiated (30 to 45 doses)
None
Unit Size:
1 L
This item is not returnable. View return policy

Product Code. 16635745

Brand: Cytiva SH30075.05

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used to enhance cell culture performance; a cost-efficient alternative to Fetal Bovine Serum (FBS).

  • Sourced from a closed herd maintained on USDA-licensed premises
  • Cattle from these closed herds are used exclusively for the production of donor bovine blood products
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Certificate of Suitability No. R1-CEP 2001-259
Country of Origin United States
Form Liquid
Product Type Adult Bovine Serum
Quantity 1 L
Sterilization Method Sequential Triple-filtered, 100 nm
Processing None
Endotoxin Concentration ≤ 50 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 25 mg/dL
Product Line HyClone™
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.