missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Defined Fetal Bovine Serum (FBS), US Origin
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 kGy)
Irradiated (30 to 45 kGy)
Irradiated, Heat Inactivated
MSC Screen, Heat Inactivated
TET Screen, Irradiated
Packungsgröße:
100mL
500mL
Beschreibung
- Assay results for over 50 components, with additional testing available
- Sourced and processed in the United States with complete traceability to origin
- Ultra-low endotoxin levels (≤ 10 EU/mL) for reduced research and production risk
- Serial 40 nm filter process removes up to 8 logs viral challenge
- Post-filtration treatments such as γ-irradiated and heat-inactivated options are also available
- Broad virus panel testing in accordance with 9 CFR 113.53
Spezifikation
Spezifikation
| Country of Origin | United States |
| Product Type | Fetal Bovine Serum |
| Quantity | 500 mL |
| Sterilization Method | Sequential-filtered, 40 nm |
| Grade | Defined |
| Processing | Irradiated (30 to 45 kGy) |
| Tested For | Virus-Tested to 9 CFR Standard |
| Volume (Metric) | 500 mL |
| Endotoxin Concentration | ≤ 10 EU/mL |
| Hemoglobin Concentration | ≤ 10 mg/dL |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur