missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ Cosmic Calf™ Serum (U.S.)

Enhance cell culture growth with U.S.-sourced HyClone™ Cosmic Calf™ Serum, a superior bovine calf serum fortified with iron and growth promoting factors for a broad spectrum of cell types.

Brand:  GE Healthcare Hyclone SH30087.03

Code : 85

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10136253

  • £98.00 / 500mL


Estimated Shipment
15-03-2018
Add to basket

Specifications

Specifications

Manufactured under cGMP and ISO 9001 certified facilities
Albumin
None
Yes
500mL
Cosmic Calf Serum (U.S.)
Iron and naturally derived components
Store at ≤-10°C.
Filtered through three sequential 100nm pore-size rated filters.
Reovirus
Videos
Documentation

Technical Documents