missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Cosmic Calf™ Serum, EMEA Tested

Product Code. 16695765
Change view
Click to view available options
Processing:
Irradiated
Non-Irradiated
Unit Size:
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Processing unitSize
16695765 Irradiated 1 L
16605775 Non-Irradiated 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16695765 Supplier Cytiva Supplier No. SH30533.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Conformation Tested According to the European Agency for the Evaluation of Medicinal Products (EMEA)

  • High-Quality Bovine Calf Serum
  • Fortified with iron and naturally derived components
  • Augmented with additional growth-promoting components to yield an exceptional bovine calf serum
  • Excellent performance with a variety of cell types
TRUSTED_SUSTAINABILITY

Specifications

Product Type Calf Bovine Serum
Quantity 1000 mL
Sterility Sterile
Sterilization Method Sequential Triple-filtered, 100 nm
Processing Irradiated
Product Line HyClone
Test Methods European Agency for the Evaluation of Medicinal Products (EMEA) Conforming Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.